missing translation for 'onlineSavingsMsg'
Learn More

CD8b.2 Rat anti-Mouse, FITC, Clone: 53-5.8, BD

Rat Monoclonal Antibody

468.00 €

Spezifikation

Antigen CD8b.2
Klon 53-5.8
Konzentration 0.5mg/mL
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 1
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15886948
Unterlagen ansehen Aktionsdetails
BD Biosciences
553040
0.5 mg
468.00 €
1 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung

Beschreibung

The 53-5.8 monoclonal antibody specifically binds to the β chain of the CD8 differentiation antigen (Ly-3.2 or Lyt-3.2) of most mouse strains, having weak reactivity with Ly-3.1 strains (e.g., AKR, C58, MRL, PL). The CD8 α and α chains (CD8a) form heterodimers with the CD8 β chain (CD8b, Ly-3, or Lyt-3) on the surface of most thymocytes. A subpopulation of mature T lymphocytes (i.e., MHC class I-restricted T cells, including most T suppressor/cytotoxic cells) expresses almost exclusively the CD8 αβ heterodimer (the α chain is absent). Subsets of γδ TCR-bearing T cells, intestinal intraepithelial lymphocytes, and dendritic cells express CD8a without CD8b. It has been suggested that the expression of the CD8a/CD8b heterodimer is restricted to T lymphocytes which matured in the thymus or in an extrathymic environment that had been influenced by thymus-initiated neuroendocrine signals. CD8 is an antigen coreceptor on the T-cell surface which interacts with MHC class I molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck (p56lck).

Spezifikation

Spezifikation

CD8b.2
0.5mg/mL
Monoclonal
Rat
Mouse
IgG1 κ
Affinity Purified
Ly-3.2; Lyt-3.2; CD8b.2
53-5.8
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only.