missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD94 Mouse anti-Human, BB700, Clone: HP-3D9, BD Horizon™
Mouse Monoclonal Antibody
Marke: BD Biosciences 566534
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
CD94 Monoclonal antibody specifically detects CD94 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD94 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
HP-3D9 | |
Mouse | |
RUO | |
Human | |
Purified |