Learn More
CD99 Monoclonal Antibody (3B2/TA8), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
145.00 € - 336.00 €
Spezifikation
Antigen | CD99 |
---|---|
Klon | 3B2/TA8 |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16331214
|
Affymetrix eBioscience
62-0997-42 |
100 Tests |
336.00 €
100 Stück |
Verfügbar ab: 13-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16321214
|
Affymetrix eBioscience
62-0997-41 |
25 Tests |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Description: The 3B2/TA8 monoclonal antibody reacts with human CD99, also known as E2 antigen and MIC2. CD99 is an approximately 32 kDa sialoglycoprotein and is expressed by all leukocytes. CD99 is involved in T cell adhesion and activation. Applications Reported: This 3B2/TA8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3B2/TA8 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD99 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P14209 | |
CD99 | |
Primary | |
Antibody |
3B2/TA8 | |
Flow Cytometry | |
Super Bright 436 | |
Mouse | |
Human | |
4267 | |
IgG2a κ | |
4° C, store in dark, DO NOT FREEZE! | |
CD99 |