missing translation for 'onlineSavingsMsg'
Learn More

CD99 Monoclonal Antibody (3B2/TA8), Super Bright™ 436, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 62-0997-41

Additional Details : Netto-Gewicht : 0.01000kg

 View more versions of this product

Product Code. 16321214

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
View alternative products »


Description

Description

Description: The 3B2/TA8 monoclonal antibody reacts with human CD99, also known as E2 antigen and MIC2. CD99 is an approximately 32 kDa sialoglycoprotein and is expressed by all leukocytes. CD99 is involved in T cell adhesion and activation. Applications Reported: This 3B2/TA8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3B2/TA8 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.

CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell addhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the haematopoietic system to T, NK and myeloid cells.
Specifications

Specifications

CD99
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P14209
Mouse
25 Tests
Primary
Human
Antibody
IgG2a κ
Flow Cytometry
3B2/TA8
Super Bright 436
CD99
CD99
Affinity Chromatography
RUO
4267
4° C, store in dark, DO NOT FREEZE!
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers