missing translation for 'onlineSavingsMsg'
Learn More

CDC16/APC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 14307-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16882884

  • 329.85 € / 150 Mikroliter

Voraussichtliches Versanddatum: 25-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.
Spezifikation

Spezifikation

CDC16/APC6
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13042, Q8R349
ANAPC6, APC6, CDC16, CDC16 homolog, CDC16Hs, Cyclosome subunit 6
Rabbit
IgG
150 μL
-20°C
Primary
290875, 69957, 8881
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CDC16
Liquid
Cdc16
CDC16/APC6 Fusion Protein Ag5427
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate