missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5RAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14740-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Neuronal CDC2-like kinase, which is involved in the regulation of neuronal differentiation, is composed of a catalytic subunit, CDK5, and an activating subunit, p25NCK5A. The protein encoded by this gene binds to p25NCK5A and therefore may be involved in neuronal differentiation. Multiple transcript variants exist for this gene, but the full-length natures of only two have been determined.Spezifikation
CDK5RAP1 | |
Polyclonal | |
Unconjugated | |
CDK5RAP1 | |
C20orf34, C42, CDK5RAP1, CDK5RAP1.3, CDK5RAP1.4, CGI 05, HSPC167 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
252827, 51654, 66971 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BTW8, Q96SZ6, Q9JLH6 | |
CDK5RAP1 | |
CDK5RAP1 Fusion Protein Ag6409 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |