missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CDX4 Antibody, R&D Systems™
Goat Polyclonal Antibody
N.V.
Spezifikation
Antigen | CDX4 |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Anwendungen | Western Blot, Immunohistochemistry |
Klassifikation | Polyclonal |
Beschreibung
CDX4 Polyclonal specifically detects CDX4 in Human samples. It is validated for Western Blot, Immunohistochemistry.Spezifikation
CDX4 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
caudal type homeo box transcription factor 4, caudal type homeobox 4, caudal type homeobox transcription factor 4, Caudal-type homeobox protein 4, homeobox protein CDX-4 | |
CDX4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
O14627 | |
1046 | |
E. coli-derived recombinant human CDX4 Met1-Thr170 Accession # O14627 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human CDX4 in direct ELISAs and Western blots. |
For Research Use Only