Learn More
Cefixime trihydrate
Product Code. 15417329
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417329 100 mg 100mg
15407329 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15417329 Supplier Thermo Scientific Alfa Aesar Supplier No. J66094.MC
 Restricted Item
Lager vorrätig
Add to basket
missing translation for 'validMainframeMsg'
A broad spectrum third generation cephalosporin | CAS: 125110-14-7 | C16H15N5O7S2 | 453.44 g/mol
A broad spectrum third generation cephalosporin
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 125110-14-7
Molecular Formula C16H15N5O7S2
Molecular Weight (g/mol) 453.44
MDL Number MFCD00865020
InChI Key OKBVVJOGVLARMR-QSWIMTSFSA-N
PubChem CID 5362065
ChEBI CHEBI:472657
IUPAC Name (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[(carboxymethoxy)imino]acetamido]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES NC1=NC(=CS1)C(=N\OCC(O)=O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O
Specifications
Type Cefixime Trihydrate
Physical Form Powder
Melting Point 218°C to 225°C
Color Yellow
Quantity 100 mg
Sensitivity Air and light sensitive
Solubility Information Soluble in water at 55;1 μg/mL
Formula Weight 507.49
Chemical Name or Material Cefixime trihydrate