missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP70 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16280-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CEP70 plays a role in the organization of both preexisting and nascent microtubules in interphase cells. During mitosis, it is required for the organization and orientation of the mitotic spindle.Spezifikation
CEP70 | |
Polyclonal | |
Unconjugated | |
CEP70 | |
BITE, centrosomal protein 70kDa, Centrosomal protein of 70 kDa, CEP70, p10 binding protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
367153, 68121, 80321 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5PQQ9, Q6IQY5, Q8NHQ1 | |
Cep70 | |
CEP70 Fusion Protein Ag9365 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |