missing translation for 'onlineSavingsMsg'
Learn More

Cetirizine Glycerol Ester Dihydrochloride (Mixture of Diastereomers), TRC

Artikelnummer. 30136927
Change view
Click to view available options
Menge:
5 mg
50 mg
Conditionnement:
5 Milligramm
50 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Menge unitSize
30136927 5 mg 5 Milligramm
17950785 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30136927 Fournisseur Toronto Research Chemicals Code fournisseur TRCC2811305MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Summenformel C24H33Cl3N2O5
Molekulargewicht (g/mol) 535.89
Synonym 2-[2-[4-[(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-acetic Acid 2,3-Dihydroxypropyl Ester Dihydrochloride
IUPAC-Name 2,3-dihydroxypropyl 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetate
SMILES ClC1=CC=C(C(C2=CC=CC=C2)N3CCN(CCOCC(OCC(O)CO)=O)CC3)C=C1

Specifications

Chemischer Name oder Material Cetirizine Glycerol Ester Dihydrochloride (Mixture of Diastereomers)
Bezeichnung der Analyten oder Bestandteile Cetirizine Glycerol Ester Dihydrochloride (Mixture of Diastereomers)
Hinweise zur Reinheitsqualität HPLC
Versandbedingung Room Temperature
InChI-Formel InChI=1S/C24H31ClN2O5/c25-21-8-6-20(7-9-21)24(19-4-2-1-3-5-19)27-12-10-26(11-13-27)14-15-31-18-23(30)32-17-22(29)16-28/h1-9,22,24,28-29H,10-18H2
Lösungstyp Neat
Industry Type Forensics
Menge 5 mg
Reinheit (%) >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.