missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHD1L Recombinant Rabbit Monoclonal Antibody (JG35-44)
GREENER_CHOICE

Artikelnummer. 17145403
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17145403 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17145403 Lieferant Invitrogen™ Lieferanten-Nr. MA534732

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Recombinant Monoclonal Antibody

Positive Control: HepG2, human tonsil tissue, human colon cancer tissue, mouse testis tissue, HepG2.

In response to DNA strand breaks, chromatin adopts a relaxed structure due to the addition of poly(ADP-ribose) (PAR) to chromatin proteins by PARP enzymes (see PARP1; MIM 173870), and this relaxation facilitates the repair of DNA damage. CHD1L interacts with PAR and has a role in chromatin relaxation following DNA damage (Ahel et al., 2009 [PubMed 19661379]).
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen CHD1L
Anwendungen Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Recombinant Monoclonal
Klon JG35-44
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gen CHD1L
Gen-Zugriffsnummer Q86WJ1
Gen-Alias 4432404A22Rik; ALC1; amplified in liver cancer 1; amplified in liver cancer protein 1; CHD1L; CHDL; chromodomain helicase DNA binding protein 1 like; chromodomain helicase DNA binding protein 1-like; chromodomain-helicase-DNA-binding protein 1-like; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 1-like; Snf2p; Unknown (protein for MGC:128792)
Gensymbole CHD1L
Wirtsspezies Rabbit
Immunogen Recombinant protein within Human CHD1L aa 680-820.
Reinigungsverfahren Protein A
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 9557
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles, store in dark
Produkttyp Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.