missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemerin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Spezifikation
Antigen | Chemerin |
---|---|
Konzentration | 0.33 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16896453
|
Proteintech
10216-1-AP-20UL |
20 μL |
140.00 €
20 Mikroliter |
Verfügbar ab: 29-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16886453
|
Proteintech
10216-1-AP-150UL |
150 μL |
399.00 €
150 Mikroliter |
Verfügbar ab: 29-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Retinoids act through ligand-dependent transcription factors, retinoid X receptor (RXRs) and retinoic acid receptors (RARs). Tazarotene-induced gene (TIG) proteins, also designated retinoic acid receptor responder proteins or RAR-responsive proteins, can be membrane bound or secreted. TIGs act as tumor suppressor genes in human cancers and are highly expressed in skin, hair follicles and endothelial cells as well as in pancreas, spleen and intestine. TIGs are activated by tazarotene and have been implicated as growth regulators that mediate the growth suppressive effects of retinoids. TIG2 is a secreted protein that is mainly expressed in epidermis, hair follicles and endothelial cells. TIG2 is inhibited in psoriatic lesions and is activated by tazarotene in skin rafts and in epidermis of psoriatic lesions.Spezifikation
Chemerin | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q99969 | |
5919 | |
Chemerin Fusion Protein Ag0261 | |
Primary | |
-20°C | |
RARRES2 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Chemerin, HP10433, RAR responsive protein TIG2, RARRES2, TIG2 | |
RARRES2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |