missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHERP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25389-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CHERP gene ontology annotations related to this gene include negative regulation of cell proliferation; positive regulation of calcineurin-NFAT signaling cascade; release of sequestered calcium ion into cytosol; RNA processing.Spezifikation
CHERP | |
Polyclonal | |
Unconjugated | |
CHERP | |
CHERP, DAN16, DAN26, ERPROT 213 21, SCAF6, SRA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10523, 27967 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CGZ0, Q8IWX8 | |
Cherp | |
CHERP Fusion Protein Ag13428 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |