Learn More
Chicken anti-Mouse IgG Fc Secondary Antibody, Invitrogen™
Chicken Polyclonal Secondary Antibody
Marke:  Invitrogen SA172026
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11565150
  • 200.00 € / 500 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
SA1-72026 detects Mouse IgG Fc fragment in mouse samples. SA1-72026 has been successfully used in ELISA and Western blot procedures. This product may be suitable for ELISA applications, but has not been tested. The SA1-72026 immunogen is from purified mouse IgG.
Thermo Scientific Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
Spezifikation
Mouse IgG Fc
Polyclonal
Unconjugated
Purified mouse IgG.
500 μg
Secondary
4° C
IgY
ELISA, Western Blot
1.0 mg/mL
Chicken
Affinity chromatography