Learn More
Chloramine-T 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30308759
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30308759 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30308759 Supplier Dr. Ehrenstorfer Supplier No. DREA11111000AL1000
 Restricted Item
Verfügbar ab: 30-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tosylchloramide Sodium
CAS 127-65-1
Analyte or Component Names Tosylchloramide Sodium
Shipping Condition +20°C
Molecular Formula C7 H7 Cl N O2 S . Na
Certifications/Compliance ISO 17034
Synonym Benzenesulfonamide, N-chloro-4-methyl-, sodium salt (1:1), Benzenesulfonamide, N-chloro-4-methyl-, sodium salt (9CI), p-Toluenesulfonamide, N-chloro-, sodium salt (8CI), Acti-chlore, Aktiven, Aktivin, Anexol, Asepto-Sol, Aseptoclean, Berkendyl, Chloralone, Chloramine-T, Chlorasan, Chloraseptine, Chlorazan, Chlorazene, Chlorazene hydrosol, Chlorazone, Chlorozone, Chlorseptol, Cloramine T, Chloramine-T, Clorina, Clorosan, Desinfect, Euclorina, Gansil, Gyneclorina, Halamid, Heliogen, Kloramin, Kloramin B, Kloramine-T, Mannolite, Mianine, Monochloramine T, Multichlor, N-Chloro-4-methylbenzenesulfonamide sodium salt, N-Chloro-p-toluenesulfonamide sodium, N-Chloro-p-toluenesulfonamide sodium salt, N-Chlorotoluenesulfonamide sodium salt, Sodium N-chloro-4-methylbenzenesulfonamide, Sodium N-chloro-p-toluenesulfonamide, Sodium chloramine T, Sodium p-toluenesulfochloramide, Sodium p-toluenesulfonchloramide, Sodium p-toluenesulfonylchloramide, Sodium tosylchloramide, Tampules, Tochlorine, Tolamine, Tosylchloramide sodium
InChI Formula InChI=1S/C7H7ClNO2S.Na/c1-6-2-4-7(5-3-6)12(10,11)9-8;/h2-5H,1H3;/q-1;+1
SMILES [Na+].Cc1ccc(cc1)S(=O)(=O)[N-]Cl
IUPAC Name sodium;chloro-(4-methylphenyl)sulfonylazanide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 227.644
Quantity 1 mL
Formula Weight 226.978 g/mol