Learn More
Chlordimeform, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Chlordimeform free base |
| CAS | 6164-98-3 |
| Summenformel | C10 H13 Cl N2 |
| Menge | 100 mg |
| Synonym | Formamidine, N'-(4-chloro-o-tolyl)-N,N-dimethyl- (7CI,8CI), N'-(4-Chloro-2-methylphenyl)-N,N-dimethylmethanimidamide, 1-(2-Methyl-4-chlorophenyl)-3,3-dimethylformamidine, Acaron, C 8514, CDM, CDM (acaricide), CIBA 8514, Chlordimeform, Chlorophenamidin, Chlorophenamidine, Chlorphenamidine, Formamidine, N,N-dimethyl-N'-(2-methyl-4-chlorophenyl)-, Fundal, Fundal 500, Galecron, N,N-Dimethyl-N'-(2-methyl-4-chlorophenyl)formamidine, N'-(2-Methyl-4-chlorophenyl)-N,N-dimethylformamidine, N'-(4-Chloro-o-tolyl)-N,N-dimethylformamidine, N'-(p-Chloro-o-tolyl)-N,N-dimethylformamidine, RS 141, Schering 36,268 |
| InChI-Formel | InChI=1S/C10H13ClN2/c1-8-6-9(11)4-5-10(8)12-7-13(2)3/h4-7H,1-3H3 |
| SMILES | CN(C)C=Nc1ccc(Cl)cc1C |
| IUPAC-Name | N'-(4-Chlor-2-methylphenyl)-N,N-dimethylmethanimidamid |
| Molekulargewicht (g/mol) | 196.68 |
| Formelmasse | 196.0767 |