missing translation for 'onlineSavingsMsg'
Learn More

Dengue Virus 2 Envelope Antibody (144), Allophycocyanin, R&D Systems™

Product Code. 18609688 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18609688 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18609688 Supplier Novus Biologicals Supplier No. NBP305828APC
 Restricted Item
Verfügbar ab: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Dengue Virus 2 Envelope Monoclonal specifically detects Dengue Virus 2 Envelope in Virus samples. It is validated for ELISA/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus 2 Envelope
Applications ELISA, Western Blot
Classification Monoclonal
Clone 144
Conjugate APC
Dilution Western Blot, ELISA
Gene Alias Dengue 2 envelope;Dengue envelope;envelope protein, Envelope protein E
Quantity 0.1 mL
Regulatory Status RUO
Product Type Antibody
Form Purified
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.