missing translation for 'onlineSavingsMsg'
Learn More

CHM 1, Tocris Bioscience™

Potent antitumor agent; inducer of apoptosis

Marke:  RD Systems (Bio-Techne) 3868/50

Weitere Details : Netto-Gewicht : 0.00050kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413734

  • 655.33 € / 50 Milligramm


Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Inducer of apoptosis; displays potent antitumor ability in human hepatocellular carcinoma. Inhibits tubulin polymerization in vitro and in vivo. Causes cell cycle arrest at G 2M phase by activation of Cdc2 kinase activity. Induces translocation of apoptosis inducing factor (AIF) from the mitochondria to nucleus. Also exhibits vascular targeting activity through upregulation of p53 and induction of death receptor (DR5)-mediated apoptosis in HUVEC cells.
Spezifikation

Chemische Identifikatoren

154554-41-3
283.258
chm-1, chm 1, chm-1 hydrate, 1,3-dioxolo 4,5-g quinolin-8 5h-one, 6-2-fluorophenyl, 6-2-fluorophenyl-2h,5h-1,3 dioxolo 4,5-g quinolin-8-one, d0b4ca, 1,5-g quinolin-8 5h-one, 6-2-fluorophenyl, 6-2-fluoro-phenyl-5h-1,3 dioxolo 4,5-g quinolin-8-one, 6-2-fluorophenyl-1,3 dioxolo 4,5-g quinolin-8 5h-one, 6-2-fluorophenyl-1,3-dioxolo 4,5-g quinolin-8 5h-one
6-(2-fluorophenyl)-5H-[1,3]dioxolo[4,5-g]quinolin-8-one
C16H10FNO3
ZMYDAPJHGNEFGQ-UHFFFAOYSA-N
375860
C1OC2=C(O1)C=C3C(=C2)C(=O)C=C(N3)C4=CC=CC=C4F

Spezifikation

CHM 1
154554-41-3
chm-1, chm 1, chm-1 hydrate, 1,3-dioxolo 4,5-g quinolin-8 5h-one, 6-2-fluorophenyl, 6-2-fluorophenyl-2h,5h-1,3 dioxolo 4,5-g quinolin-8-one, d0b4ca, 1,5-g quinolin-8 5h-one, 6-2-fluorophenyl, 6-2-fluoro-phenyl-5h-1,3 dioxolo 4,5-g quinolin-8-one, 6-2-fluorophenyl-1,3 dioxolo 4,5-g quinolin-8 5h-one, 6-2-fluorophenyl-1,3-dioxolo 4,5-g quinolin-8 5h-one
C1OC2=C(O1)C=C3C(=C2)C(=O)C=C(N3)C4=CC=CC=C4F
283.258
283.25
50mg
C16H10FNO3
ZMYDAPJHGNEFGQ-UHFFFAOYSA-N
6-(2-fluorophenyl)-5H-[1,3]dioxolo[4,5-g]quinolin-8-one
375860
>99%
Videos