Learn More
Thermo Scientific Chemicals Cholic acid, 97%
Product Code. 10163540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10163540 100 g Glass bottle 100g
10506603 25 g Glass bottle 25g
10338943 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10163540 Supplier Thermo Scientific Chemicals Supplier No. 159111000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Lager vorrätig
Add to basket
missing translation for 'validMainframeMsg'
CAS: 81-25-4 | C24H40O5 | 408.579 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 81-25-4
Molecular Formula C24H40O5
MDL Number MFCD00003672
InChI Key BHQCQFFYRZLCQQ-OELDTZBJSA-N
Synonym cholic acid, cholalic acid, cholate, cholalin, colalin, cholsaeure, cholbam, cholicacid, cholic acid, 5beta, 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanic acid
PubChem CID 221493
ChEBI CHEBI:16359
Specifications
Melting Point 197°C to 202°C
Color White to Beige
Loss on Drying 1% max. (135°C)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 g
Beilstein 10, IV, 2072
Merck Index 15, 2210
Specific Rotation (20°C, 589nm) (c=2, ethanol)%gt; +35°
SMILES CC(CCC(=O)O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
IUPAC Name (4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molecular Weight (g/mol) 408.579
Formula Weight 408.57
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material Cholic acid