missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CHR 2797
Aminopeptidase inhibitor
N.V.
Spezifikation
Reinheit | >98% |
---|---|
Gen-Alias | Tosedostat |
Produkttyp | CHR 2797 |
Molekulargewicht | 406.48 |
Hemmstoffe | CHR 2797 |
Beschreibung
Aminopeptidase inhibitor (IC50 values are 100, 150, 220, > 1000, > 5000, > 10000 and > 30000 nM for LAP, PuSA, aminopeptidase N, aminopeptidase B, PILSAP, LTA4 hydrolase and MetAP2 respectively). Potently inhibits tumor cell proliferation in a variety of tumor cell lines in vitro and in vivo.Spezifikation
>98% | |
CHR 2797 | |
CHR 2797 |
Tosedostat | |
406.48 |
For Research Use Only