Learn More
Thermo Scientific Chromeleon™ 7.4 License Code - New
Product Code. 31906281 Shop All Thermo Scientific Products
Change view
Click to view available options
Operating System:
Windows 11
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Operating System Unit Size
31906281 Windows 11 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31906281 Supplier Thermo Scientific Supplier No. 7450.0104A
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
This license code is required for the activation of a new Chromeleon 7.4 installation; adding a license to an existing Chromeleon installation (7.4.x or lower) may require different license P/N.
Includes:
  • License code
  • License certificate
Note: The latest minor version of Chromeleon 7.4.x will be provided. This will be a digital delivery of Chromeleon (download option). No installation media is provided.
TRUSTED_SUSTAINABILITY
Specifications
Type Chromatography Software
For Use With (Equipment) Chromeleon CDS
Certifications/Compliance 21 CFR
Product Line Chromeleon 7
Operating System Windows 11
Quantity 1 Ea.