Learn More
Thermo Scientific™ Chromeleon™ CDS Single Edition (SE) License
Product Code. 31903479 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Chromeleon CDS
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31903479 Chromeleon CDS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31903479 Supplier Thermo Scientific™ Supplier No. 7100.0108
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
Single Edition (SE) Chromeleon™ CDS license enabling control of one Thermo Scientific instrument with data acquisition, processing, and reporting capabilities.
  • Chromeleon™ CDS Single Edition (SE) license for control of one Thermo Scientific instrument
  • Includes one client enabling local or remote instrument control
  • Supports instrument control, data acquisition, processing, and reporting
  • Includes Instrument Controller and Class 1 instrument license
  • Includes Data Client, Instrument Operation, and Report Designer Pro
  • Includes Fraction Collection functionality
  • SE license is a stand-alone license and cannot be added to an existing Chromeleon installation
  • Non-Thermo Scientific instruments cannot be added to this configuration
  • License must be ordered with the appropriate license key or license code depending on Chromeleon version
TRUSTED_SUSTAINABILITY
Specifications
Type Data System
For Use With (Equipment) Chromeleon CDS