missing translation for 'onlineSavingsMsg'
Learn More

CIAO1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 10295-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16886583

  • 329.85 € / 150 Mikroliter

Verfügbar ab
16-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

10295-1-AP detected 48-55 kDa band because of phosphorylation. 10295-1-AP does well in IHC and IF detection.

WDR39 is a member of the WD40 family of proteins. WDR39 specifically interacts with WT1 both in vitro and in vivo. This interaction results in a decrease in transcriptional activation mediated by WT1. WDR39 does not inhibit binding of WT1 to its consensus nucleotide sequence and does not affect the repression activity of WT1. Thus, WDR39 appears to specifically modulate the transactivation activity of WT1 and may function to regulate the physiological functions of WT1 in cell growth and differentiation.
Spezifikation

Spezifikation

CIAO1
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O76071
CIA1, CIAO1, WDR39
Rabbit
IgG
150 μL
-20°C
Primary
9391
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CIAO1
Liquid
CIAO1
CIAO1 Fusion Protein Ag0349
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation