missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cidofovir-Diphosphat-Trinatriumsalz (> 85%), TRC
Specifications
Specifications
| Chemischer Name oder Material | Cidofovir Diphosphate Trisodium Salt |
| CAS | 352525-57-6 |
| Summenformel | C8 H13 N3 Na3 O12 P3 |
| Menge | 1 mg |
| Synonym | Diphosphoric acid Monoanhydride with [[(1S)-2-(4-Amino-2-oxo-1(2H)-pyrimidinyl)-1-(hydroxymethyl)ethoxy]methyl]phosphonic Acid Trisodium Salt |
| InChI-Formel | InChI=1S/C8H16N3O12P3.3Na/c9-7-1-2-11(8(13)10-7)3-6(4-12)21-5-24(14,15)22-26(19,20)23-25(16,17)18;;;/h1-2,6,12H,3-5H2,(H,14,15)(H,19,20)(H2,9,10,13)(H2,16,17,18);;;/q;3*+1/p-3/t6-;;;/m0.../s1 |
| SMILES | NC1=NC(=O)N(C[C@@H](CO)OCP(=O)(O[Na])OP(=O)(O[Na])OP(=O)(O)O[Na])C=C1 |
| IUPAC-Name | [[(2S)-1-(4-Amino-2-oxopyrimidin-1-yl)-3-hydroxypropan-2-yl]oxymethyl-[[hydroxy(nadiooxy)phosphoryl]oxy-nadiooxyphosphoryl]oxyphosphoryl]oxynatrium |
| Molekulargewicht (g/mol) | 505.092 |
| Formelmasse | 504.941 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?