missing translation for 'onlineSavingsMsg'
Learn More

cis-ACBD, Tocris Bioscience™

Potent, selective L-glutamate uptake inhibitor

Marke:  Tocris (Bio-Techne) 027150

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13417523

  • 509.62 € / 50 Milligramm


Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Potent, competitive and selective inhibitor of glutamate uptake. Certain confusion exists over the naming of this compound because of apparent contradictions in the literature. This is the isomer which has the carboxylic acid and the amino groups on the same side of the cyclobutyl ring.
Spezifikation

Chemische Identifikatoren

73550-55-7
159.141
2,4-methanoglutamate, cis-acbd, 2,4-methanoglutamic acid, 1s,3s-1-aminocyclobutane-1,3-dicarboxylic acid, cis-1-amino-1,3-cyclobutanedicarboxylic acid, trans-1-aminocyclobutane-1,3-dicarboxylic acid, cis-1-aminocyclobutane-1,3-dicarboxylic acid, cis-1-aminocyclobutane-1,3-dicarboxylic acid hydrate, trans-1-aminocyclobutane-1,3-dicarboxylic acid hydrate, 1,3-cyclobutanedicarboxylic acid, 1-amino-, cis-, hydrate
1-aminocyclobutane-1,3-dicarboxylic acid
C6H9NO4
GGMYWPBNZXRMME-UHFFFAOYSA-N
1214
C1C(CC1(C(=O)O)N)C(=O)O

Spezifikation

cis-ACBD
73550-55-7
2,4-methanoglutamate, cis-acbd, 2,4-methanoglutamic acid, 1s,3s-1-aminocyclobutane-1,3-dicarboxylic acid, cis-1-amino-1,3-cyclobutanedicarboxylic acid, trans-1-aminocyclobutane-1,3-dicarboxylic acid, cis-1-aminocyclobutane-1,3-dicarboxylic acid, cis-1-aminocyclobutane-1,3-dicarboxylic acid hydrate, trans-1-aminocyclobutane-1,3-dicarboxylic acid hydrate, 1,3-cyclobutanedicarboxylic acid, 1-amino-, cis-, hydrate
C1C(CC1(C(=O)O)N)C(=O)O
159.141
159.14
50mg
C6H9NO4
GGMYWPBNZXRMME-UHFFFAOYSA-N
1-aminocyclobutane-1,3-dicarboxylic acid
1214
>98%
Videos