missing translation for 'onlineSavingsMsg'
Learn More

cis-Decahydro-2a,4a,6a,8a-tetraazacyclopent[fg]acenaphthylen, Thermo Scientific Chemicals

Catalog Number p-7029361
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
79236-92-3
C10H18N4
194.282
MFCD09263319
YSPZOYMEWUTYDA-UHFFFAOYSA-N
cis-glyoxal-cyclen, 2a,4a,6a,8a-decahydrotetraazacyclopent fg acenaphthylene, cis-decahydro-2a,4a,6a,8a-tetraazacyclopent fg acenaphthylene, cyclen cis-glyoxal derivative, 1,1':3,3'-diethano-2,2'-biimidazolidine, decahydro-2a,4a,6a,8a-tetraazacyclopenta fg acenaphthylene, cis-decahydro-2a,4a,6a,8a-tetraazacyclopent fg acena, 1,4,7,10-tetraazatetracyclo-5,5,2,04,13,010,14 tetradecane, 1,4,7,10-tetraazatetracyclo-5,5,2,04.13,010.14 tetradecane, 4aalpha,8abeta-1,8:4,5-diethanodecahydro-1,4,5,8-tetraazanaphthalene
9794169
C1CN2CCN3CCN4C3C2N1CC4
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
79236-92-3
C10H18N4
194.282
MFCD09263319
YSPZOYMEWUTYDA-UHFFFAOYSA-N
cis-glyoxal-cyclen, 2a,4a,6a,8a-decahydrotetraazacyclopent fg acenaphthylene, cis-decahydro-2a,4a,6a,8a-tetraazacyclopent fg acenaphthylene, cyclen cis-glyoxal derivative, 1,1':3,3'-diethano-2,2'-biimidazolidine, decahydro-2a,4a,6a,8a-tetraazacyclopenta fg acenaphthylene, cis-decahydro-2a,4a,6a,8a-tetraazacyclopent fg acena, 1,4,7,10-tetraazatetracyclo-5,5,2,04,13,010,14 tetradecane, 1,4,7,10-tetraazatetracyclo-5,5,2,04.13,010.14 tetradecane, 4aalpha,8abeta-1,8:4,5-diethanodecahydro-1,4,5,8-tetraazanaphthalene
9794169
C1CN2CCN3CCN4C3C2N1CC4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material cis-Decahydro-2a,4a,6a,8a-tetraazacyclopent[fg]acenaphthylene
Schmelzpunkt 29°C to 31°C
Menge 250 mg
Empfindlichkeit Hygroscopic
Formelmasse 194.28

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'