missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse STAT4 Antibody, R&D Systems™
Description
STAT4 Polyclonal specifically detects STAT4 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
Specifications
| Antigen | STAT4 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000 dilution, Immunoprecipitation 3 ug/500 ug cell lysate |
| Gene Alias | signal transducer and activator of transcription 4, SLEB11 |
| Gene Symbols | STAT4 |
| Host Species | Rabbit |
| Immunogen | aa 730 - 749 of murine STAT4 |
| Quantity | 200 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?