Learn More
Greiner Bio-One μClear™ 96-well, Cell Repellent-Treated, Bioprinting Kit - V-Shaped-Bottom Microplate
Artikelnummer. 15810715
Change view
Click to view available options
Farbe:
Klar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Farbe
15810715 Klar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15810715 Lieferant Greiner Bio-One Lieferanten-Nr. 781841
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Farbe:Transparent; Sterilität:Nicht steril; Deckel:Mit Deckel
  • 3D in a 2D workflow.
  • Scalable technology with different size/well-number plate systems available.
  • Compatible with fluorescence microscopy, western blotting, qRT-PCR, flow cytometry, viability assays, chemiluminescence, etc.
  • The core technology is the magnetization of cells with biocompatible NanoShuttle™-PL.
  • The reproducible formation of one spheroid per well in an F-bottom plate with Cell-Repellent surface is forced by magnets to form structurally and biologically representative 3D models in-vitro.
  • Magnetized cells in a plate are printed into spheroids by placing atop a drive of magnets.
  • One magnet below each well uses mild magnetic forces to induce cell aggregation and print one spheroid at the bottom of each well anywhere within 15 minutes to a few hours.
  • Afterwards, the spheroids can be cultured long-term without the use of magnetic force.
  • This system overcomes the limitations of other platforms by enabling rapid formation of spheroids reproducible and scalable in size for high-throughput formats (96, 384 and 1536 well) and without limitation to cell types.
  • The 3D printing method together with commercially available standardized biochemical assay methods to facilitate continuous assessment of cell viability and other functions, provides an ideal combination for high-throughput compound screening.
  • Adding and removing solutions is made easy by the use of magnetic forces to hold down spheroids during aspiration limiting spheroid loss.
  • Spheroids can also be picked up and transferred between vessels using magnetic tools such as the MagPen™.
  • Magnetic forces can also be used to create co-cultures with fine spatial organization.
TRUSTED_SUSTAINABILITY
Spezifikation
Format Standard
Deckel Mit Deckel
Array 16 x 24
Farbe Klar
Anzahl Wells 96
Sterilität Unsteril
Oberflächenbehandlung Zellabweisend
Wellform Konischer Boden (V-Well)
Enthält 3 x 600μL vials of NanoShuttle™-PL, 6-well levitating drive, 96-well spheroid drive, 96-well holding drive, 96-well ring drive, 96-well deep well plate, 2 x 6-well culture multiwell plates with cell-repellent surface, 2 x 96-well cell culture microplates with cell0repellent surface
Anzahl pro Packung 5/Pk.
Menge 5/Pk.