Learn More
Greiner Bio-One μClear™ 96-well, Cell Repellent-Treated, Bioprinting Kit - V-Shaped-Bottom Microplate
Beschreibung
- 3D in a 2D workflow.
- Scalable technology with different size/well-number plate systems available.
- Compatible with fluorescence microscopy, western blotting, qRT-PCR, flow cytometry, viability assays, chemiluminescence, etc.
- The core technology is the magnetization of cells with biocompatible NanoShuttle™-PL.
- The reproducible formation of one spheroid per well in an F-bottom plate with Cell-Repellent surface is forced by magnets to form structurally and biologically representative 3D models in-vitro.
- Magnetized cells in a plate are printed into spheroids by placing atop a drive of magnets.
- One magnet below each well uses mild magnetic forces to induce cell aggregation and print one spheroid at the bottom of each well anywhere within 15 minutes to a few hours.
- Afterwards, the spheroids can be cultured long-term without the use of magnetic force.
- This system overcomes the limitations of other platforms by enabling rapid formation of spheroids reproducible and scalable in size for high-throughput formats (96, 384 and 1536 well) and without limitation to cell types.
- The 3D printing method together with commercially available standardized biochemical assay methods to facilitate continuous assessment of cell viability and other functions, provides an ideal combination for high-throughput compound screening.
- Adding and removing solutions is made easy by the use of magnetic forces to hold down spheroids during aspiration limiting spheroid loss.
- Spheroids can also be picked up and transferred between vessels using magnetic tools such as the MagPen™.
- Magnetic forces can also be used to create co-cultures with fine spatial organization.
Spezifikation
Spezifikation
| Format | Standard |
| Deckel | Mit Deckel |
| Array | 16 x 24 |
| Farbe | Klar |
| Anzahl Wells | 96 |
| Sterilität | Unsteril |
| Oberflächenbehandlung | Zellabweisend |
| Wellform | Konischer Boden (V-Well) |
| Enthält | 3 x 600μL vials of NanoShuttle™-PL, 6-well levitating drive, 96-well spheroid drive, 96-well holding drive, 96-well ring drive, 96-well deep well plate, 2 x 6-well culture multiwell plates with cell-repellent surface, 2 x 96-well cell culture microplates with cell0repellent surface |
| Anzahl pro Packung | 5/Pk. |
| Mehr anzeigen |