Learn More
Cloxacillin Benzathine (2:1), TRC
Artikelnummer. 30137671
Change view
Click to view available options
Menge:
100 mg
200 mg
1 g
Packungsgröße:
1 Gramm
100 Milligramm
200 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30137671 200 mg 200 Milligramm
30133203 1 g 1 Gramm
30005796 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137671 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCC587465200MG
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 23736-58-5
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[[3-(2-chlorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-, (2S,5R,6R)-, compd. with N,N'-bis(phenylmethyl)-1,2-ethanediamine (2:1)∣4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[3-(o-chlorophenyl)-5-methyl-4-isoxazolecarboxamido]-3,3-dimethyl-7-oxo-, compd. with N,N'-dibenzylethylenediamine (2:1) (8CI)∣4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[[3-(2-chlorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-, [2S-(2α,5α,6β)]-, compd. with N,N'-bis(phenylmethyl)-1,2-ethanediamine (2:1)∣1,2-Ethanediamine, N,N'-bis(phenylmethyl)-, bis[(2S,5R,6R)-6-[[[3-(2-chlorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-1-carboxylate] (9CI)∣1,2-Ethanediamine, N,N'-bis(phenylmethyl)-, bis[[2S-(2α,5α,6β)]-6-[[[3-(2-chlorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-1-carboxylate]∣Ethylenediamine, N,N'-dibenzyl-, bis[6-[3-(o-chlorophenyl)-5-methyl-4-isoxazolecarboxamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-1-carboxylate] (8CI)∣Ethylenediamine, N,N'-dibenzyl-, bis[6-[3-(o-chlorophenyl)-5-methyl-4-isoxazolecarboxamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate] (8CI)∣Benzathine cloxacillin∣Boviclox∣Cloxacillin benzathin∣Cloxacillin benzathine∣Dry-Clox∣Noroclox DC∣Opticlox∣Orbenin Dry Cow∣Triclox
Spezifikation
Format Neat
Summenformel C54H56Cl2N8O10S2
Molekulargewicht 1112.11
InChI-Schlüssel COCFKSXGORCFOW-VZHMHXRYSA-N
SMILES C(CNCC1=CC=CC=C1)NCC1=CC=CC=C1.CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C(O)=O)C(=NO1)C1=CC=CC=C1Cl.CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C(O)=O)C(=NO1)C1=CC=CC=C1Cl
IUPAC-Name bis((2S,5R,6R)-6-[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-amido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid); benzyl[2-(benzylamino)ethyl]amine
Produkttyp API
Menge 200 mg
Molekulargewicht (g/mol) 1112.11
Versandbedingung Room Temperature