missing translation for 'onlineSavingsMsg'
Learn More

Clozapine N-oxide, 99%, Thermo Scientific Chemicals

Product Code. 18936930
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18936930 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18936930 Supplier Thermo Scientific Alfa Aesar Supplier No. J67235.LB0
 Restricted Item
Verfügbar ab: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 34233-69-7
Color Yellow
Molecular Formula C18H19ClN4O
MDL Number MFCD00210190
Quantity 5 mg
UN Number UN2811
Synonym clozapine n-oxide, clozapine-n-oxide, unii-mza8bk588j, chembl1688, mza8bk588j, n-oxyclozapine, clozapine n-oxide cno, oguczbiqsyywef-uhfffaoysa-n
Solubility Information Soluble in DMSO.
InChI Key WYRDWWAASBTJLM-UHFFFAOYSA-N
SMILES C[N+]1(CCN(CC1)C2=C3C=CC=CC3=NC4=C(N2)C=C(C=C4)Cl)[O-]
IUPAC Name 3-chloro-6-(4-methyl-4-oxidopiperazin-4-ium-1-yl)-5H-benzo[b][1,4]benzodiazepine
Molecular Weight (g/mol) 342.827
PubChem CID 2819
Formula Weight 342.83
Percent Purity 99%
Physical Form Solid
Chemical Name or Material Clozapine N-oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.