missing translation for 'onlineSavingsMsg'
Learn More

Clozapine, Tocris Bioscience™

Dopamine antagonist with some D4 selectivity. Also 5-HT2A/2C antagonist

Marke:  Tocris (Bio-Techne) 0444/500

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 500mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13417663

  • 831.96 € / 500 Milligramm


Voraussichtlicher Liefertermin 22-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Atypical antipsychotic drug, with a much lower tendency to cause extrapyramidal side effects than conventional neuroleptics. Displays a broad range of pharmacological actions; the antipsychotic effects are thought to be mediated principally by 5-HT2A/2C and dopamine receptor blockade (Ki values are 21, 170, 170, 230 and 330 nM for D4, D3, D1, D2 and D5 receptors respectively).
Spezifikation

Chemische Identifikatoren

5786-21-0
326.828
clozapine, leponex, clozapin, clozaril, fazaclo, clorazil, iprox, clozapinum, asaleptin, clozapina
3-chloro-6-(4-methylpiperazin-1-yl)-5H-benzo[b][1,4]benzodiazepine
C18H19ClN4
ZUXABONWMNSFBN-UHFFFAOYSA-N
2818
CN1CCN(CC1)C2=C3C=CC=CC3=NC4=C(N2)C=C(C=C4)Cl

Spezifikation

Clozapine
5786-21-0
clozapine, leponex, clozapin, clozaril, fazaclo, clorazil, iprox, clozapinum, asaleptin, clozapina
CN1CCN(CC1)C2=C3C=CC=CC3=NC4=C(N2)C=C(C=C4)Cl
326.828
326.83
500mg
C18H19ClN4
ZUXABONWMNSFBN-UHFFFAOYSA-N
3-chloro-6-(4-methylpiperazin-1-yl)-5H-benzo[b][1,4]benzodiazepine
2818
>99%
Videos