missing translation for 'onlineSavingsMsg'
Learn More

CMV UL147/vCXC2 Mouse anti-Viral, Clone: 146326, R&D Systems™

Mouse Monoclonal Antibody

N.V.

Spezifikation

Antigen UL147/vCXC2
Klon 146326
Immunogen E. coli-derived recombinant viral CMV UL147/vCXC2
Gen-ID (Entrez) 3077419
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

UL147/vCXC2 Monoclonal specifically detects UL147/vCXC2 in Viral samples. It is validated for Western Blot.
Spezifikation

Spezifikation

UL147/vCXC2
E. coli-derived recombinant viral CMV UL147/vCXC2
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Primary
Detects viral CMV UL147/vCXC2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant viral (rv) CMV UL146, rvMCV type 2, rvMIP-I, rvMIP-II, or rvMIP-III is observed.
Unconjugated
Western Blot
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
146326
3077419
Reconstitute at 0.5 mg/mL in sterile PBS.
Mouse
IgG2a
Protein A or G purified from hybridoma culture supernatant
RUO
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only