Learn More
COL6A5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560254
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MDVVFLIDNS RNIAKDEFKA VKALVSSVID NFNIASDPLI SDSGDRIALL SYSPWESSRR KMGTVKTEFD FITYDNQLLM KNHIQTSFQQ LNGEATIGRA L Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 69%. Immunohistochemistry retrieval method: HIER pH 6.
This gene encodes a member of the collagen superfamily of proteins. The encoded protein contains multiple von Willebrand factor A-like domains and may interact with the alpha 1 and alpha 2 chains of collagen VI to form the complete collagen VI trimer. Polymorphisms in this gene may be linked to dermal phenotypes, such as eczema. Alternative splicing results in multiple transcript variants.Spezifikation
COL6A5 | |
Polyclonal | |
Unconjugated | |
COL6A5 | |
Col29a1; Col6a5; Collagen alpha-1(XXIX) chain; collagen alpha-5(VI) chain; collagen type VI alpha 5; collagen type VI alpha 5 chain; collagen, type VI, alpha 5; collagen, type XXIX, alpha 1; EG665033; Gm7455; RGD1565804; von Willebrand factor A domain-containing protein 4; VWA4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
256076 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A8TX70 | |
COL6A5 | |
Recombinant protein corresponding to Human COL6A5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only