Specifications
| Quantity | 1 |
| Material | PTFE |
| For Use With (Equipment) | Standard Taper Ground Glass Joints |
| Description | Fisherbrand™ Colored PTFE Conical Joint Clamp |
| Size | Joint 34/45 |
| Color | Orange |