missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

N.V.

Spezifikation

Antigen Complement Component C2
Immunogen Mouse myeloma cell line NS0-derived recombinant human Complement Component C2 Ala21-Leu752 Accession # P06681
Gen-ID (Entrez) 717
Gen-Zugriffsnummer P06681
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

Complement Component C2 Polyclonal specifically detects Complement Component C2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Spezifikation

Spezifikation

Complement Component C2
717
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Primary
Detects human Complement Component C2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) C1r, rhC1s, rhC3a, and rhC5a is observed.
Unconjugated
Western Blot, Immunoprecipitation
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Mouse myeloma cell line NS0-derived recombinant human Complement Component C2 Ala21-Leu752 Accession # P06681
P06681
Reconstitute at 0.2 mg/mL in sterile PBS.
Goat
IgG
Affinity Purified
RUO
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only