missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COMT Polyclonal Antibody

Artikelnummer. 16344545 Alle ansehen Thermo Scientific Produkte
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16344545 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16344545 Lieferant Invitrogen™ Lieferanten-Nr. PA595276

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Human COMT shares 81.9% and 81% amino acid (aa) sequence identity with mouse and rat COMT, respectively. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human SH-SY5Y whole cell, human HepG2 whole cell, human A431 whole cell, human MCF-7 whole cell, rat liver tissue. IHC: Mouse Lung tissue, Rat Lung tissue, Human Placenta tissue IHC-F: human placenta tissue, mouse lung tissue, rat lung tissue. ICC/IF: A549 Cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Catechol-O-methyltransferase catalyzes the transfer of a methyl group from S-adenosylmethionine to catecholamines, including the neurotransmitters dopamine, epinephrine, and norepinephrine. This O-methylation results in one of the major degradative pathways of the catecholamine transmitters. In addition to its role in the metabolism of endogenous substances, COMT is important in the metabolism of catechol drugs used in the treatment of hypertension, asthma, and Parkinson disease. COMT is found in two forms in tissues, a soluble form (S-COMT) and a membrane-bound form (MB-COMT). The differences between S-COMT and MB-COMT reside within the N-termini. Several transcript variants are formed through the use of alternative translation initiation sites and promoters.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen COMT
Anwendungen Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konzentration 500 μg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 5mg BSA and 0.05mg sodium azide
Gen Comt
Gen-Zugriffsnummer O88587, P21964, P22734
Gen-Alias Catechol O-methyltransferase; catecholamine-O-methyltransferase; catechol-O-methyltransferase; catechol-O-methyltransferase 1; catechol-O-methyltransferase isoform; Comt; Comt1; D16Wsu103e; D330014B15Rik; epididymis secretory sperm binding protein Li 98n; HEL-S-98n; OTTHUMP00000197749; OTTHUMP00000197750; testicular tissue protein Li 42
Gensymbole Comt
Wirtsspezies Rabbit
Immunogen E.coli-derived human COMT recombinant protein (Position: G52-P271).
Reinigungsverfahren Affinity chromatography
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 12846, 1312, 24267
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Lyophilized
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.