missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin-32 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10450-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.Spezifikation
Connexin-32 | |
Polyclonal | |
Unconjugated | |
GJB1 | |
CMTX, CMTX1, Connexin-32, CX32, Gap junction beta 1 protein, GJB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14618, 2705 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08034, P28230 | |
Gjb1 | |
Connexin-32 Fusion Protein Ag0659 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |