Learn More
Decon™ Contrad 70 Liquid Detergent
Product Code. 15937922
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15937922 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15937922 Supplier Decon™ Supplier No. CON705
 Restricted Item
Lager vorrätig
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Concentrated liquid detergent. Ideal for laboratory washing by hand, for soaking or for use with an ultrasonic bath. For removing soil, grit, organic contaminants, inorganic residue, chemicals, and solvents from glass, ferrous metal, ceramic and plastics.
  • Phosphate- and chlorine-free
  • Free-rinsing
  • Specifications
    Form Liquid
    Volume (Metric) 5 L
    compliance-icons compliance-icons
    Product Identifier
    • Contrad 70
    Signal Word
    • Danger
    Hazard Category
    • Acute toxicity Category 4
    • Serious eye damage/eye irritation Category 1
    • Skin corrosion/irritation Category 2
    Hazard Statement
    • H302-Harmful if swallowed.
    • H315-Causes skin irritation.
    • H319-Causes serious eye irritation.
    Precautionary Statement
    • P284-[In case of inadequate ventilation] wear respiratory protection.
    • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
    • P305-IF IN EYES:Separate eyelids with finger tips..
    • P351-Rinse cautiously with water for several minutes.
    • P361-Take off immediately all contaminated clothing.
    Supplemental information
    • MIXTURE LIST-Contain: Potassium hydroxide,Trisodium citrate dihydrate