missing translation for 'onlineSavingsMsg'
Learn More

Kupfer(I)-oxid, 97 % (Cu + Cu2O Assay), Thermo Scientific Chemicals

Catalog Number p-4407430
Click to view available options
Menge:
500 g
2 kg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1317-39-1
Cu2O
MFCD00010974
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1317-39-1
Cu2O
MFCD00010974

Rotes Pigment für Glas- und Keramikglasuren und als Katalysator

Red pigment for glass and ceramic glazes, and as a catalystCopper(I) oxide is used in antifouling, marine paint, brazing paste, antifungal treatment for plant seeds. Used as a hydrogen scavenger in copper brass and bronze and high grade lubricants. Finds uses in glass and ceramic pigments, ferrite magnets, catalyst in organic synthesis, dyes and dye intermediates and as catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Siedepunkt 1800°C (decomposition)
Chemischer Name oder Material Copper(I) oxide
Schmelzpunkt 1235°C
Namenshinweis Cu + Cu2O Assay
Farbe Violett bis Rot
Physikalische Form Pulver
Menge 500 g
Prozentgehaltsbereich 97%
Löslichkeitsinformationen Practically insoluble in water. Soluble in NH4OHInsoluble in water but soluble in acids.
Formelmasse 143.08
Geruch Odorless
Brechungsindex 2.705
Empfindlichkeit Air and moisture sensitive
Aussehen Purple-red powder
Dichte 6 g/cm3
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'