missing translation for 'onlineSavingsMsg'
Learn More

Kupferkugel, 99.99 % (Metallbasis), frei von Sauerstoff, Thermo Scientific Chemicals

Catalog Number p-4408720
Click to view available options
Menge:
500 g
2 kg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
Kupfer
[Cu]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
Kupfer
[Cu]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Copper
Prozentgehaltsbereich 99.99% (metals basis)
Menge 500 g
Form Shot

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'