missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CLTCL1 Mouse Monoclonal Antibody, Proteintech
Alle ansehen Proteintech ProdukteDescription
The CLTCL1 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human CLTCL1. This antibody recognizes human antigen. The CLTCL1 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
| Antigen | CLTCL1 |
| Anwendungen | Immunocytochemistry/Immunofluorescence |
| Klon | 1B5A10 |
| Konzentration | 2000 μg/mL |
| Konjugat | CoraLite Plus 488 |
| Gensymbole | CHC22; CLH22; CLTCL; CLTD; FLJ36032 |
| Wirtsspezies | Mouse |
| Immunogen | Peptide |
| Reinigungsverfahren | Protein A purification |
| Menge | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?