Learn More
CoraLite™ Plus 488-conjugated HSP60 Rabbit Polyclonal Antibody, Proteintech
Alle ansehen Proteintech ProdukteBeschreibung
The HSP60 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human HSP60. This antibody recognizes human, mouse, rat antigen. The HSP60 antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.
Spezifikation
Spezifikation
| Antigen | HSP60 |
| Anwendungen | Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Konzentration | 1000 μg/mL |
| Konjugat | CoraLite Plus 488 |
| Gensymbole | CPN60; GROEL; HLD4; HSP60; HSP65; HuCHA60; SPG13 |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant Protein |
| Reinigungsverfahren | Antigen affinity purification |
| Menge | 20 μL |
| Primär oder sekundär | Primary |
| Mehr anzeigen |