missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10611-2-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Defects in COX10 are a cause of mitochondrial complex IV deficiency (MT-C4D); also known as cytochrome c oxidase deficiency. A disorder of the mitochondrial respiratory chain with heterogeneous clinical manifestations, ranging from isolated myopathy to severe multisystem disease affecting several tissues and organs. Features include hypertrophic cardiomyopathy, hepatomegaly and liver dysfunction, hypotonia, muscle weakness, excercise intolerance, developmental delay, delayed motor development and mental retardation. A subset of patients manifest Leigh syndrome.Spezifikation
COX10 | |
Polyclonal | |
Unconjugated | |
COX10 | |
COX10, Heme O synthase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1352, 691853, 70383 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q12887, Q8CFY5 | |
Cox10 | |
COX10 Fusion Protein Ag0930 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |