missing translation for 'onlineSavingsMsg'
Learn More

CPNE1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 10126-2-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16886303

  • 112.71 € / 20 Mikroliter

Voraussichtliches Versanddatum: 18-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene encodes a calcium-dependent protein that also contains two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. However, the encoded protein does not contain a predicted signal sequence or transmembrane domains. This protein has a broad tissue distribution and it may function in membrane trafficking. This gene and the gene for RNA binding motif protein 12 overlap at map location 20q11. 21. Alternate splicing results in multiple transcript variants encoding different proteins.
Spezifikation

Spezifikation

CPNE1
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q99829
Copine 1, copine I, COPN1, CPN1, CPNE1
Rabbit
IgG
20 μL
-20°C
Primary
8904
Western Blot
Unconjugated
CPNE1
Liquid
CPNE1
CPNE1 Fusion Protein Ag0139
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation