missing translation for 'onlineSavingsMsg'
Learn More

CPS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 24513-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16882985

  • 112.71 € / 20 Mikroliter

Voraussichtliches Versanddatum: 18-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The mitochondrial enzyme encoded by this gene catalyzes synthesis of carbamoyl phosphate from ammonia and bicarbonate. This reaction is the first committed step of the urea cycle, which is important in the removal of excess urea from cells. The encoded protein may also represent a core mitochondrial nucleoid protein. Three transcript variants encoding different isoforms have been found for this gene. The shortest isoform may not be localized to the mitochondrion. Mutations in this gene have been associated with carbamoyl phosphate synthetase deficiency, susceptibility to persistent pulmonary hypertension, and susceptibility to venoocclusive disease after bone marrow transplantation.
Spezifikation

Spezifikation

CPS1
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P07756, P31327, Q8C196
CPS1, CPSase I
Rabbit
IgG
20 μL
-20°C
Primary
1373, 227231, 497840
Western Blot
Unconjugated
CPS1
Liquid
Cps1
CPS1 Fusion Protein Ag21605
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation