missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREB5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14196-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The product of this gene belongs to the CRE (cAMP response element)-binding protein family. Members of this family contain zinc-finger and bZIP DNA-binding domains. The encoded protein specifically binds to CRE as a homodimer or a heterodimer with c-Jun or CRE-BP1, and functions as a CRE-dependent trans-activator. Alternatively spliced transcript variants encoding different isoforms have been identified.Spezifikation
CREB5 | |
Polyclonal | |
Unconjugated | |
CREB5 | |
CRE BPA, CREB 5, CREB5, CREBPA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231991, 500131, 9586 | |
-20°C | |
Liquid |
Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q02930, Q8K1L0 | |
Creb5 | |
CREB5 Fusion Protein Ag5399 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |