Learn More
Tocris Bioscience™ Cryptotanshinone
STAT3 inhibitor. Also displays multiple other activities
Marke:  Tocris Bioscience™ 371350
Weitere Details : Netto-Gewicht : 0.01000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13413474
  • 546.02 € / 50 Milligramm
Voraussichtlicher Liefertermin 25-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Major tanshinone isolated from Salvia miltiorrhiza that exhibits multiple activities. Exhibits antitumor activity via inhibition STAT3 activity (IC 50 = 4.6 μM). Displays antibacterial and anti-inflammatory activity and acts as an antidiabetes and antiobesity agent via activation of AMP-activated protein kinase (AMPK). Also improves cognitive impairment in Alzheimer's disease transgenic mice by inhibition of acetylcholinesterase (IC 50 = 4.09 μM) and reduction in Aβ peptide generation.
Spezifikation
Spezifikation
Cryptotanshinone
>98
50mg
Cryptotanshinone
296.36