missing translation for 'onlineSavingsMsg'
Learn More

CSM-ARG-URA (Powder), MP Biomedicals™

Product Code. 15796375 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
15796375 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15796375 Supplier MP Biomedicals Supplier No. 114523112
  • ONLINE EXCLUSIVE PRICE
    104.00 €
    98.70 € / 10 Gramm
    Sparen 5.30 €
    5% Rabatt
 Restricted Item
Verfügbar ab: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Complete supplement mixture (without arginine or uracil) for richer medium, increased yield/growth/probability of successful plasmid formations.

  • Single dropout formulation (without arginine and uracil).
  • The selection of plasmids in yeast is based on the use of auxotrophic mutant strains that cannot grow without a specific media component (an amino acid, purine or pyrimidine).
  • Transformation with a plasmid containing the mutated gene enables the transformant to grow on a medium lacking the required component.
  • This CSM-Dropout medium creates a richer medium for better yield and growth rate, and increases the probability of successful transformations when screening libraries or performing gene knock-outs.
  • For use with DOB/DOBA or YNB with the appropriate nitrogen and carbon sources to custom formulate your own selective synthetic defined yeast media.
  • Research Use Only (RUO).

Specifications

Color Colorless to faint yellow
CAS 73-24-5
pH 4.0 to 4.8
Quantity 10 g
Physical Form Powder
Solubility Information Soluble in water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.