Learn More
CTLA-4 Antibody (CTLA4/6868R), Novus Biologicals™
Artikelnummer. 18666324 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μg
20 μg
Packungsgröße:
100 Mikrogramm
20 Mikrogramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18666324 100 μg 100 Mikrogramm
18658464 20 μg 20 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18666324 Lieferant Novus Biologicals Lieferanten-Nr. NBP320470100ug
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Monoclonal Antibody
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen CTLA-4
Anwendungen Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon CTLA4/6868R
Konzentration 0.2 mg/ml
Konjugat Unconjugated
Verdünnung Immunohistochemistry-Paraffin 1-2 μg/mL
Zusammensetzung 10mM PBS with 0.05% BSA
Gen-Alias CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Wirtsspezies Rabbit
Immunogen Recombinant fragment of human CTLA-4 (exact sequence is proprietary)
Reinigungsverfahren Protein A or G purified
Menge 100 μg
Regulatorischer Status RUO
Forschungsgebiet Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction
Primär oder sekundär Primary
Gen-ID (Entrez) 1493
Zielspezies Human
Inhalt und Lagerung Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG κ