missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR7 Mouse, BV421, Clone: 10D1, BD Horizon™
Mouse Monoclonal Antibody
Marke: BD Biosciences 566234
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
CXCR7 Monoclonal antibody specifically detects CXCR7 in Human, Mouse samples. It is validated for Flow cytometry.
CXCR7 Monoclonal antibody specifically detects CXCR7 in Human, Mouse samples. It is validated for Flow cytometry.Spezifikation
CXCR7 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 μg | |
Cell Biology | |
Human, Mouse | |
Purified |
Flow Cytometry | |
10D1 | |
Brilliant Violet 421 | |
ACKR3, CMKOR, RDC1, GPR159 | |
Human CXCR7 Transfected Cell Line | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
IgG2a κ |
For Research Use Only.